Natriumacetat trihydrat ≥99%
Leverantör: Thermo Fisher Scientific
A16230.0C A16230.0E A16230.36
A16230.0CEA 6900 SEK
A16230.0C A16230.0E A16230.36
Natriumacetat trihydrat ≥99%
Natriumacetat trihydrat
Purity: 99% CAS No.: 6131-90-4 Molecular Formula: NaOOCCH3.3H2O Formula Weight: 136.08 (82.03anhy) Odour: Odourless pH: 7.5-9.0 Melting Point: 62-64° Beilstein Reference: 3732037 Merck Reference: 14,8571 MDL No.: MFCD00071557
Formel: H₃CCOONa·3H₂O
Kokpunkt: 120…123 °C (1013 hPa)
Smältpunkt: 62…64 °C
Densitet: 1,45
Storage Temperature: Omgivande
MDL Number: MFCD00071557
CAS nummer: 6131-90-4
EINECS: 204-823-8
Merck Index: 14,8571
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=313c1d45673fab192565649f8cbb7ddff555de39&vid=1ad37a966605626c95117735c83b16eaea381fbc&prid=web&clid=SAPDAM