Sodium acetate hydrate ≥99.9985%, Puratronic®
Leverantör: Thermo Fisher Scientific
41685.06 41685.14 41685.22
41685.06EA 1475 SEK
41685.06 41685.14 41685.22
Sodium acetate hydrate ≥99.9985%, Puratronic®
Sodium acetate hydrate
Purity: 99.9985% CAS No.: 41484-91-7 Molecular Formula: NaOOCCH3.xH2O (x~3) Formula Weight: 82.03(anhy) Physical Form: Powder Density: 1.45 Melting Point: 58° Merck Reference: 14,8571 MDL No.: MFCD00071557
Formel: H₃CCOONa·3H₂O
MW: 100.05 g/mol
Smältpunkt: 58 °C
Densitet: 1.45
Storage Temperature: Omgivande
MDL Number: MFCD00071557
CAS nummer: 41484-91-7
EINECS: 204-823-8
Merck Index: 14,8571