N-2-aminoetyl-1-naftylamin dihydroklorid, Supelco®
Leverantör: Merck
Varning
Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine
M-Clarity™ quality level = MQ100
70720-100G 70720-25G
70720-100GEA 9450 SEK
70720-100G 70720-25G
N-2-aminoetyl-1-naftylamin dihydroklorid, Supelco®
N-2-aminoetyl-1-naftylamin dihydroklorid
Formel: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Smältpunkt: 188…190 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012556
CAS nummer: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a3a8caa6eebed857ff4c23fe6c26f60dfaa65a80&vid=826a0c6f1f36376a44faea35b53f684bea8d57ee&prid=web&clid=SAPDAM