Guanidiniumtiocyanat, Millipore®
Leverantör: Merck
Fara
Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
M-Clarity™ quality level = MQ100
1.04167.0250 1.04167.1000
1.04167.0250EA 2075 SEK
1.04167.0250 1.04167.1000
Guanidiniumtiocyanat, Millipore®
Guanidiniumtiocyanat
Formel: CH₅N₃·CHNS
MW: 118,16 g/mol
Smältpunkt: 118 °C
Densitet: 1,29 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013027
CAS nummer: 593-84-0
EINECS: 209-812-1
UN: 1759
ADR: 8,III