Guanidiniumtiocyanat ≥99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Guanidinium sulphocyanide, GTC, Guanidine Thiocyanate, Guanidinium rhodanide
B21250.0E B21250.22 B21250.36
B21250.0EEA 20025 SEK
B21250.0E B21250.22 B21250.36
Guanidiniumtiocyanat ≥99%
Guanidiniumtiocyanat
Purity: 99% CAS No.: 593-84-0 Molecular Formula: CH5N3.HSCN Formula Weight: 118.16 Melting Point: 118-122° Beilstein Reference: 3563461
Formel: NH₂C(=NH)NH₂·HSCN
MW: 118.16 g/mol
Smältpunkt: 118…122 °C
Densitet: 1.29
Storage Temperature: Omgivande
MDL Number: MFCD00013027
CAS nummer: 593-84-0
EINECS: 209-812-1