Guanidinhydroklorid, Millipore®
Leverantör: Merck
Varning
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
M-Clarity™ quality level = MQ100
1.04220.0100 1.04220.1000 1.04220.5000 1.04220.9025
1.04220.0100EA 657 SEK
1.04220.0100 1.04220.1000 1.04220.5000 1.04220.9025
Guanidinhydroklorid, Millipore®
Guanidinhydroklorid
Formel: NH₂C(=NH)NH₂·HCl
Smältpunkt: 185 °C
Densitet: 1,354 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM