Guanidinhydroklorid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
A13543.0I A13543.0B A13543.30
A13543.0IEA 6675 SEK
A13543.0I A13543.0B A13543.30
Guanidinhydroklorid ≥98%
Guanidinhydroklorid
Purity: 98% CAS No.: 50-01-1 Molecular Formula: CH5N3.HCl Formula Weight: 95.53 Melting Point: 178-185° Beilstein Reference: 3591990 Merck Reference: 14,4562 MDL No.: MFCD00013026
Formel: NH₂C(=NH)NH₂·HCl
MW: 95,53 g/mol
Smältpunkt: 178…185 °C
Densitet: 1,344
Storage Temperature: Omgivande
MDL Number: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 14,4562
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=ab8b4c12372c0550614e1f8ab87bea02ff8530c3&prid=web&clid=SAPDAM