4-(Methylamino)phenol sulphate 99%
Leverantör: Thermo Fisher Scientific
Varning
A16236.0B A16236.18 A16236.30
A16236.0BEA 4675 SEK
A16236.0B A16236.18 A16236.30
4-(Methylamino)phenol sulphate 99%
4-(Methylamino)phenol sulphate
Purity: 99% CAS No.: 55-55-0 Molecular Formula: C14H18N2O2.H2SO4 Formula Weight: 344.38 Melting Point: ca 260° dec. Beilstein Reference: 3919382 Merck Reference: 14,6017 UN No.: 2811 MDL No.: MFCD00013140
Formel: C₁₄H₂₀N₂O₆S
Kokpunkt: 227 °C (1013 hPa)
Smältpunkt: ∼ 260 °C (dec.)
Densitet: 1,434 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013140
CAS nummer: 55-55-0
EINECS: 200-237-1
UN: 2811
ADR: 6.1,III
Merck Index: 14,6017