Tris-(hydroximetyl) aminometan hydroklorid
Leverantör: RESEARCH ORGANICS
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
RES3098T-B103X RES3098T-B101X RES3098T-B102X
RESERES3098T-B103XEA 0 SEK
RESERES3098T-B103X RESERES3098T-B101X RESERES3098T-B102X
Tris-(hydroximetyl) aminometan hydroklorid
Tris-(hydroximetyl) aminometan hydroklorid
Formel: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Smältpunkt: 150…152 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012590
CAS nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM