Tris-(hydroximetyl) aminometan hydroklorid 99
Leverantör: Thermo Fisher Scientific
Synonyms: Tris(hydroxymethyl)methylamine hydrochloride, 2-Amino-2-(hydroxymethyl)propane-1,3-diol hydrochloride, Trizma® hydrochloride, Trizma(R) hydrochloride, Tris(hydroxymethyl)aminomethane hydrochloride, TRIS HCl, Tromethane hydrochloride, Tris-HCl, Tris(hydroxymethyl)methylammonium chloride, Trometamol hydrochloride, THAM HCl
A11379.0B A11379.18 A11379.30
A11379.0BEA 6700 SEK
A11379.0B A11379.18 A11379.30
Tris-(hydroximetyl) aminometan hydroklorid 99
Tris-(hydroximetyl) aminometan hydroklorid
Purity: 99+% CAS No.: 1185-53-1 Molecular Formula: C4H11NO3.HCl Formula Weight: 157.60 pH: 3.5-5.0 Melting Point: ca 152° dec. Beilstein Reference: 3675235 MDL No.: MFCD00012590
Formel: NH₂C(CH₂OH)₃·HCl
MW: 157.6 g/mol
Smältpunkt: ∼ 152 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00012590
CAS nummer: 1185-53-1
EINECS: 214-684-5
Merck Index: 12,09902
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cbdc9446fdb9bcfe3f1d992ff20a16726f98113&vid=3d668801395b81fa595401a752279006da8fd3cb&prid=web&clid=SAPDAM