2,3,5-Trifenyltetrazoliumklorid ≥98%, Sigma-Aldrich®
Leverantör: SIGMA ALDRICH MICROSCOPY
Varning
Synonyms: Tetrazolium salt, Tetrazolium Red, 2,3,5-Triphenyl-2H-tetrazol-3-ium chloride, 2,3,5-Triphenyl-2H-tetrazolium chloride, TTC
M-Clarity™ quality level = MQ200
T8877-100G T8877-5G T8877-10G T8877-25G T8877-50G
T8877-100GEA 5300 SEK
T8877-100G T8877-5G T8877-10G T8877-25G T8877-50G
2,3,5-Trifenyltetrazoliumklorid ≥98%, Sigma-Aldrich®
2,3,5-Trifenyltetrazoliumklorid
Formel: C₁₉H₁₅ClN₄
MW: 334.81 g/mol
Kokpunkt: ∼360 °C (1013 hPa)
Smältpunkt: 235
Storage Temperature: Kylskåp
MDL Number: MFCD00011963
CAS nummer: 298-96-4
EINECS: 206-071-6
Merck Index: 13,09815
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c333c35973a3c23046a4cf82f13618dcc1c7d9ac&vid=64898dcfe1e696e6f65ede885a0d2331e18e3b01&prid=web&clid=SAPDAM