Kaliumhydroxid concentrate for dilution to 0.5 N i vattenlösning, Acculute™ standard volumetric solution
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Potash caustic, Caustic potash
88602.LQ 88602.LS
88602.LQEA 1450 SEK
88602.LQ 88602.LS
Kaliumhydroxid concentrate for dilution to 0.5 N i vattenlösning, Acculute™ standard volumetric solution
Kaliumhydroxid
CAS No.: 1310-58-3 Molecular Formula: KOH Formula Weight: 56.11 Physical Form: Liquid Appearance: Colourless UN No.: 1814 MDL No.: MFCD00003553
Formel: KOH
MW: 56.11 g/mol
Kokpunkt: 1320
Smältpunkt: 360
Storage Temperature: Omgivande
MDL Number: MFCD00003553
CAS nummer: 1310-58-3
EINECS: 215-181-3
UN: 1814
ADR: 8 II
Merck Index: 12,07806