L (+)-Histidine-monohydroklorid-monohydrat
Leverantör: Molekula
Synonyms: L-Histidine monohydrochloride monohydrate, (2S)-(+)-Histidine monohydrochloride monohydrate, L-Histidine hydrochloride monohydrate, (S)-(+)-Histidine monohydrochloride monohydrate, His HCl monohydrate, H-His-OH.HCl.H2O, H-His-OH·HCl·H₂O, L-3-Imidazol-4-ylalanine monohydrochloride monohydrate, L-Histidine hydrochloride hydrate
41915465-1KG 41915465-100G
MOLE41915465-1KGEA 0 SEK
MOLE41915465-1KG MOLE41915465-100G
L (+)-Histidine-monohydroklorid-monohydrat
L (+)-Histidine-monohydroklorid-monohydrat
Formel: C₆H₉N₃O₂·HCl·H₂O
Smältpunkt: 259 °C
Densitet: 1,485 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00151027
CAS nummer: 5934-29-2
EINECS: 211-438-9
Merck Index: 13,04741
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=561d2381f930afe618f78d680790ea9a43f6596c&vid=8aacf6dc4ef5382410cb743498272b48c3c36999&prid=web&clid=SAPDAM