L (+)-Cysteinhydrokloridmonohydrat, Millipore®
Leverantör: Merck
Varning
Synonyms: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
M-Clarity™ quality level = MQ100
1.02839.0025 1.02839.0100 1.02839.1000 1.02839.9010
1.02839.0025EA 479 SEK
1.02839.0025 1.02839.0100 1.02839.1000 1.02839.9010
L (+)-Cysteinhydrokloridmonohydrat, Millipore®
L (+)-Cysteinhydrokloridmonohydrat
Formel: HSCH₂CH(NH₂)COOH·HCl·H₂O
MW: 175,64 g/mol
Smältpunkt: 168…170 °C
Storage Temperature: Omgivande
MDL Number: MFCD00065606
CAS nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM