Tetrabutylammonium hydroxide 40% (w/w) i vattenlösning
Leverantör: Thermo Fisher Scientific
Fara
L02809.18 L02809.30 L02809.0B
L02809.18EA 995 SEK
L02809.18 L02809.30 L02809.0B
Tetrabutylammonium hydroxide 40% (w/w) i vattenlösning
Tetrabutylammonium hydroxide
CAS No.: 2052-49-5 Molecular Formula: C16H37NO Formula Weight: 259.48 Flash Point: None Refractive Index: 1.4080 Odour: Amine-like pH: 14.0 Beilstein Reference: 3571228 UN No.: 3267 MDL No.: MFCD00009425
Formel: C₁₆H₃₇NO
MW: 259.48 g/mol
Smältpunkt: 29.85
Densitet: 0.990
Storage Temperature: Omgivande
MDL Number: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3267
ADR: 8 II