Rubidium hydroxide 50% (w/w) i vattenlösning , 99.6%
Leverantör: Thermo Fisher Scientific
Fara
10565.NC 10565.NE
10565.NCEA 2750 SEK
10565.NC 10565.NE
Rubidium hydroxide 50% (w/w) i vattenlösning , 99.6%
Rubidium hydroxide
Purity: 99.6+% CAS No.: 1310-82-3 Molecular Formula: HORb Formula Weight: 102.48(anhy) Physical Form: Liquid Appearance: Pale yellow Merck Reference: 14,8285 UN No.: 2677 MDL No.: MFCD00011194
Formel: RbOH
MW: 102.48 g/mol
Smältpunkt: 300…302 °C
Densitet: 1.74 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00011194
CAS nummer: 1310-82-3
EINECS: 215-186-0
UN: 2677
ADR: 8 II
Merck Index: 14,8285