Selenic acid 40% i vattenlösning
Leverantör: Thermo Fisher Scientific
Fara
18851.22 18851.36
18851.22EA 6575 SEK
18851.22 18851.36
Selenic acid 40% i vattenlösning
Selenic acid
CAS No.: 7783-08-6 Molecular Formula: H2O4Se Formula Weight: 144.97(anhy) Physical Form: Liquid Density: 1.41 Appearance: Colourless Merck Reference: 14,8429 UN No.: 2922 MDL No.: MFCD00012191
Formel: H₂O₄Se
MW: 144.97
Kokpunkt: 260
Smältpunkt: 58
Densitet: 1.41
Storage Temperature: Omgivande
MDL Number: MFCD00012191
CAS nummer: 7783-08-6
EINECS: 231-979-4
UN: 2922
ADR: 8 II
Merck Index: 14,8429