Silvernitrat solution_standardised 1.0 N aqueous solution
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
35597.AP 35597.K2
35597.APEA 16425 SEK
35597.AP 35597.K2
Silvernitrat solution_standardised 1.0 N aqueous solution
Silvernitrat
CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless UN No.: 3264 MDL No.: MFCD00003414
Formel: AgNO₃
MW: 169.87 g/mol
Kokpunkt: 444
Smältpunkt: 210
Storage Temperature: Omgivande
MDL Number: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 3264
ADR: 8 II
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM