Silvernitrat standardised solution 0.1 N i vattenlösning
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
35601.AP 35601.K2 35601.M4
35601.APEA 1525 SEK
35601.AP 35601.K2 35601.M4
Silvernitrat standardised solution 0.1 N i vattenlösning
Silvernitrat
CAS No.: 7761-88-8 Molecular Formula: AgNO3 Formula Weight: 169.87 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Colourless Odour: Odourless UN No.: 3082 MDL No.: MFCD00003414
Formel: AgNO₃
MW: 169.87 g/mol
Kokpunkt: 444
Smältpunkt: 210
Storage Temperature: Omgivande
MDL Number: MFCD00003414
CAS nummer: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM