Tetrabutylammonium hydroxide 1 M i metanol
Leverantör: Thermo Fisher Scientific
Fara
212911000 212918000 212910025
ACRO212911000EA 1500 SEK
ACRO212911000 ACRO212918000 ACRO212910025
Tetrabutylammonium hydroxide 1 M i metanol
Tetrabutylammonium hydroxide
Formel: C₁₆H₃₇NO
Smältpunkt: –98 °C
Densitet: 0,83 g/cm³
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009425
CAS nummer: 2052-49-5
EINECS: 218-147-6
UN: 3286
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Solution
Titration with HCl 0.95 to 1.10 M