Bariumklorid solution_standardised 0.5 N aqueous solution
Leverantör: Thermo Fisher Scientific
Varning
35583.K2 35583.K7
35583.K2EA 2075 SEK
35583.K2 35583.K7
Bariumklorid solution_standardised 0.5 N aqueous solution
Bariumklorid
CAS No.: 10361-37-2 Molecular Formula: BaCl2 Formula Weight: 208.25 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00003445
Formel: BaCl₂
MW: 208.24 g/mol
Kokpunkt: 1560
Smältpunkt: 963
Densitet: 1.034 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003445
CAS nummer: 10361-37-2
EINECS: 233-788-1
UN: 3287
ADR: 6.1 III
Merck Index: 12,00998