Natriumhydroxid 0.1 M NaOH (0.1N) 99 Volumetric, Fluka™
Leverantör: Honeywell Chemicals
Varning
Synonyms: Natronlut, Kaustik soda
319481-2L 319481-500ML 319481-6X500ML 319481-1L 319481-2.5L
319481-2LEA 975 SEK
319481-2L 319481-500ML 319481-6X500ML 319481-1L 319481-2.5L
Natriumhydroxid 0.1 M NaOH (0.1N) 99 Volumetric, Fluka™
Natriumhydroxid
Formel: NaOH
MW: 40 g/mol
Kokpunkt: 100 °C (1.013 hPa)
Smältpunkt: 323
Densitet: 1.000 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00003548
CAS nummer: 1310-73-2
EINECS: 231-791-2
UN: 1824
ADR: 8 III
Merck Index: 14,08627
Order Now
Specifikation testresultat
Assay titr. 0.095 - 0.105 mol/l
Carbonate (CO₃) Max. 0.0015 mol/l
Traceable to NIST SRM effective
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective