Oxalsyra 5% w/v aqueous solution
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ethanedioic acid
35619.K2 35619.K7
35619.K2EA 1400 SEK
35619.K2 35619.K7
Oxalsyra 5% w/v aqueous solution
Oxalsyra
Molecular Formula: HO2CCO2H Formula Weight: 90.04 Physical Form: Liquid Odour: Odourless UN No.: 3265 MDL No.: MFCD00002573
Formel: C₂H₂O₄
MW: 90.04 g/mol
Kokpunkt: 365
Storage Temperature: Omgivande
MDL Number: MFCD00002573
CAS nummer: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8 III
Merck Index: 13,06980