Saltsyra 1 mol/l, EMPROVE® EXPERT BiopharmaceuticalProduction, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Varning
Synonyms: Hydrochloric acid solution, Muriatic acid
1.10165.9025 1.10165.2500 1.10165.9190
1.10165.9025EA 0 SEK
1.10165.9025 1.10165.2500 1.10165.9190
Saltsyra 1 mol/l, EMPROVE® EXPERT BiopharmaceuticalProduction, SAFC®
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8