Saltsyra 0.5 mol/l (0.5 N) Volumetric
Leverantör: REAGECON
Varning
Synonyms: Hydrochloric acid solution, Muriatic acid
H20505 H20501
REGEH20505EA 870 SEK
REGEH20505 REGEH20501
Saltsyra 0.5 mol/l (0.5 N) Volumetric
Saltsyra
Formel: HCl
MW: 36.46 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II