Saltsyra 0.2 mol/l (0.2 N) Volumetric
Leverantör: REAGECON
Synonyms:
Hydrochloric acid solution, Muriatic acid
H20205
H20201
REGEH20205EA
1625
SEK
REGEH20205
REGEH20201
Saltsyra 0.2 mol/l (0.2 N) Volumetric
Saltsyra
Formel:
HCl MW: 36.46 g/mol Densitet: 1.004 g/cm³ (25 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00011324 CAS nummer: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8 III |