EDTA disodium salt 0.1 M EDTA-Na2 i vattenlösning Reag. Ph. Eur. volumetrisk lösning, for complexometry, Fluka™
Leverantör: Honeywell Chemicals
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
34550-10L-VP 34550-1L 34550-6X1L 34550-500ML 34550-2.5L
34550-10L-VPEA 5025 SEK
34550-10L-VP 34550-1L 34550-6X1L 34550-500ML 34550-2.5L
EDTA disodium salt 0.1 M EDTA-Na2 i vattenlösning Reag. Ph. Eur. volumetrisk lösning, for complexometry, Fluka™
EDTA disodium salt
Formel: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Densitet: 1.01 g/cm³
Storage Temperature: Omgivande
MDL Number: MFCD00070672
CAS nummer: 139-33-3
EINECS: 382-266-9
Merck Index: 12,03556
Order Now
Specifikation testresultat
Factor 0.999 - 1.001
Traceable to NIST SRM effective