Glutaraldehyd 50% i vattenlösning
Leverantör: Thermo Fisher Scientific
|
Fara
|
CAS No.: 111-30-8
Molecular Formula: C5H8O2
Formula Weight: 100.12
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Density: 1.123
Refractive Index: 1.4210
Appearance: Colourless to pale yellow
Odour: Stench
Melting Point: ca -21°
Boiling Point: 100°
Beilstein Reference: 605390
Merck Reference: 14,4472
UN No.: 2922
MDL No.: MFCD00007025
Formel:
C₅H₈O₂ Kokpunkt: 100 °C Smältpunkt: ∼–21 °C Densitet: 1,123 Storage Temperature: Kylskåp |
MDL Number:
MFCD00007025 CAS nummer: 111-30-8 EINECS: 203-856-5 UN: 2922 ADR: 8,III Merck Index: 14,4472 |