Glutaraldehyd 25% i vattenlösning
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Pentandial
A17876.AP A17876.0F A17876.AE
A17876.APEA 680 SEK
A17876.AP A17876.0F A17876.AE
Glutaraldehyd 25% i vattenlösning
Glutaraldehyd
CAS No.: 111-30-8 Molecular Formula: C5H8O2 Formula Weight: 100.12 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.062 Refractive Index: 1.3730 Appearance: Colourless Beilstein Reference: 605390 Merck Reference: 14,4472 UN No.: 2922 MDL No.: MFCD00007025
Formel: C₅H₈O₂
MW: 100.12 g/mol
Kokpunkt: 188
Smältpunkt: –6 °C
Densitet: 1.062
Flampunkt: >71 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00007025
CAS nummer: 111-30-8
EINECS: 203-856-5
UN: 2922
ADR: 8 III
Merck Index: 14,4472