(±)-2-Methylpiperazine 98%
Leverantör: Thermo Scientific
|
Fara
|
Synonyms:
2-Methylpiperazine
127471000
127475000
127470025
ACRO127471000EA
925
SEK
ACRO127471000
ACRO127475000
ACRO127470025
(±)-2-Methylpiperazine 98%
(±)-2-Methylpiperazine
Formel:
C₅H₁₂N₂ MW: 100,16 g/mol Kokpunkt: 154…156 °C (760 mmHg) Smältpunkt: 61…66 °C Flampunkt: 63 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00005954 CAS nummer: 109-07-9 EINECS: 203-644-2 UN: 2925 ADR: 4.1,II |
Specifikation testresultat
Appearance | White to yellow Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Melting point | 61 to 66 °C |
Water | ≤1 % |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...