(±)-1-Penten-3-ol 97%
Leverantör: Thermo Fisher Scientific
Varning
130010050 130010250 130011000
ACRO130010050EA 370 SEK
ACRO130010050 ACRO130010250 ACRO130011000
(±)-1-Penten-3-ol 97%
(±)-1-Penten-3-ol
Formel: C₅H₁₀O
Kokpunkt: 114…115 °C (760 mmHg)
Densitet: 0,839 g/cm³
Flampunkt: 25 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004573
CAS nummer: 616-25-1
EINECS: 210-472-1
UN: 1987
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4220 to 1.4260 (20°C, 589 nm)
GC ≥96.0 %