(±)-1-Hepten-3-ol ≥98%
Leverantör: Thermo Fisher Scientific
Varning
A14651.09 A14651.18
A14651.09EA 1050 SEK
A14651.09 A14651.18
(±)-1-Hepten-3-ol ≥98%
(±)-1-Hepten-3-ol
Purity: 98% CAS No.: 4938-52-7 Molecular Formula: C7H14O Formula Weight: 114.18 Flash Point: 54°(129°F) Refractive Index: 1.4330 Boiling Point: 154-156° Beilstein Reference: 1720340 UN No.: 1987 MDL No.: MFCD00021940
Formel: C₇H₁₄O
Kokpunkt: 154…156 °C
Densitet: 0,836
Flampunkt: 54 °C (129 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00021940
CAS nummer: 4938-52-7
EINECS: 225-579-9
UN: 1987
ADR: 3,III