(±)-3-Buten-2-ol 97%
Leverantör: Thermo Scientific
Fara
Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol
387070250 387070050
ACRO387070250EA 1875 SEK
ACRO387070250 ACRO387070050
(±)-3-Buten-2-ol 97%
(±)-3-Buten-2-ol
Formel: C₄H₈O
MW: 72,11 g/mol
Kokpunkt: 96…98 °C (760 mmHg)
Smältpunkt: –100 °C
Densitet: 0,83 g/cm³
Flampunkt: 16 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004543
CAS nummer: 598-32-3
EINECS: 209-929-8
UN: 1987
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4130 to 1.4170 (20°C, 589 nm)
GC ≥96.0 %