N-Acetyl-DL-tryptophan ≥99%
Leverantör: Thermo Fisher Scientific
Synonyms: Ac-DL-Trp-OH
A17562.14 A17562.22
A17562.14EA 900 SEK
A17562.14 A17562.22
N-Acetyl-DL-tryptophan ≥99%
N-Acetyl-DL-tryptophan
Purity: 99% CAS No.: 87-32-1 Molecular Formula: C13H14N2O3 Formula Weight: 246.27 Melting Point: ca 205° dec. Beilstein Reference: 89478
Formel: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
Smältpunkt: ∼ 205 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00005644
CAS nummer: 87-32-1
EINECS: 201-739-3