N-Acetyl-DL-tryptophan 98+%, extra pure
Leverantör: Thermo Fisher Scientific
Synonyms: Ac-DL-Trp-OH
227150250 227151000
ACRO227150250EA 550 SEK
ACRO227150250 ACRO227151000
N-Acetyl-DL-tryptophan 98+%, extra pure
N-Acetyl-DL-tryptophan
Formel: C₁₃H₁₄N₂O₃
MW: 246.27 g/mol
Smältpunkt: 204…206 °C (dec.)
Storage Temperature: Kylskåp
MDL Number: MFCD00005644
CAS nummer: 87-32-1
EINECS: 201-739-3
Order Now
Specifikation testresultat
Appearance White to beige Crystalline powder or crystals
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C)
Sulfated ash ≤0.2 %
Titration excess NaOH 98.0 to 101.0 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm