5-Bromo-DL-tryptophan 99%
Leverantör: Thermo Fisher Scientific
Synonyms: 2-Amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
107510010 107515000 107510050
ACRO107510010EA 3250 SEK
ACRO107510010 ACRO107515000 ACRO107510050
5-Bromo-DL-tryptophan 99%
5-Bromo-DL-tryptophan
Formel: C₁₁H₁₁BrN₂O₂
MW: 283.12 g/mol
Smältpunkt: 261…267 °C (dec.)
Storage Temperature: Frys
MDL Number: MFCD00005648
CAS nummer: 6548-09-0
EINECS: 229-464-4
Order Now
Specifikation testresultat
Appearance White to light beige to light grey Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Titration with HClO4 98.5 to 101.5 %