(±)-4-Metyl-2-pentanol ≥99%
Leverantör: Thermo Scientific
Varning
Synonyms: (±)-Methyl isobutyl carbinol
A13435.AP A13435.0F
A13435.APEA 505 SEK
A13435.AP A13435.0F
(±)-4-Metyl-2-pentanol ≥99%
(±)-4-Metyl-2-pentanol
Purity: 99% CAS No.: 108-11-2 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 41°(105°F) Refractive Index: 1.4110 Odour: Slightly sweetish Melting Point: -90° Boiling Point: 131-133° Beilstein Reference: 1098268 UN No.: 2053 MDL No.: MFCD00004550
Formel: C₆H₁₄O
MW: 102,18 g/mol
Kokpunkt: 131…133 °C
Smältpunkt: –90 °C
Densitet: 0,806
Flampunkt: 41 °C (105 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00004550
CAS nummer: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3,III