(±)-2-Methyl-1-butanol 98%
Leverantör: Thermo Scientific
Varning
158270010 158270025 158270250 158271000
ACRO158270010EA 1025 SEK
ACRO158270010 ACRO158270025 ACRO158270250 ACRO158271000
(±)-2-Methyl-1-butanol 98%
(±)-2-Methyl-1-butanol
Formel: C₅H₁₂O
MW: 88,15 g/mol
Kokpunkt: 129 °C (1013 hPa)
Smältpunkt: –70 °C
Densitet: 0,81 g/cm³
Flampunkt: 43 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004743
CAS nummer: 137-32-6
EINECS: 205-289-9
UN: 1105
ADR: 3,III
Merck Index: 12,06109
Order Now
Specifikation testresultat
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4090 to 1.4120 (20°C, 589 nm)
GC ≥97.5 %
Specific optical rotation -0.5° to +0.5° (20°C, 589 nm) (Neat)
Sulfate (SO4) ≤0.05 %