(±)-2-Methylbutyric acid ≥97.0%
Leverantör: TCI EUROPE
Fara
Synonyms: (±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid
M0181-25ML M0181-500ML
TCIAM0181-25MLEA 215 SEK
TCIAM0181-25ML TCIAM0181-500ML
(±)-2-Methylbutyric acid ≥97.0%
(±)-2-Methylbutyric acid
Formel: C₅H₁₀O₂
MW: 102,13 g/mol
Kokpunkt: 174°C
Smältpunkt: –70 °C
Densitet: 0,938 g/cm³ (20 °C)
Flampunkt: 73°C
Storage Temperature: Omgivande
MDL Number: MFCD00002669
CAS nummer: 116-53-0
EINECS: 204-145-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39989b049c18333c8b8c3fa56ece63f47a08a300&vid=39989b049c18333c8b8c3fa56ece63f47a08a300&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 95.0 %