3-Methyl-2-cyclopenten-1-one 98% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: 3-Methylcyclopent-2-enone, 3-Methyl-2-cyclopentenone
163590100 163590500 163592500
ACRO163590100EA 1325 SEK
ACRO163590100 ACRO163590500 ACRO163592500
3-Methyl-2-cyclopenten-1-one 98% stabilised
3-Methyl-2-cyclopenten-1-one
Formel: C₆H₈O
MW: 96,13 g/mol
Kokpunkt: 74 °C (15 mmHg)
Smältpunkt: 3…5 °C
Densitet: 0,98 g/cm³
Flampunkt: 65 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001403
CAS nummer: 2758-18-1
EINECS: 220-421-5
Order Now
Specifikation testresultat
Appearance Clear light yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4870 to 1.4900 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer 0.1 % hydroquinone
UV at 240 nm A: ≤0.26
Wavelength at 1 A.U. ≤215 nm
Residue after evaporation ≤2 ppm
Acidity ≤0.0002 meq/g
Alkalinity ≤0.0006 meq/g
Peroxides ≤0.0075 %