1,3-Cyclohexadiene 96% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Cyclohexa-1,3-diene
111100050 111100250 111101000
ACRO111100050EA 1225 SEK
ACRO111100050 ACRO111100250 ACRO111101000
1,3-Cyclohexadiene 96% stabilised
1,3-Cyclohexadiene
Formel: C₆H₈
MW: 80,13 g/mol
Kokpunkt: 80 °C (1013 hPa)
Smältpunkt: –98 °C
Densitet: 0,86 g/cm³
Flampunkt: –6 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00001532
CAS nummer: 592-57-4
EINECS: 209-764-1
UN: 3295
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4715 to 1.4765 (20°C, nm)
GC ≥95.0 %
Stabilizer 0.1 % BHT