1-Bromo-2-iodobenzene 99% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Bromo-1-iodobenzene, 2-Bromoiodobenzene
221950050 221950250
ACRO221950050EA 685 SEK
ACRO221950050 ACRO221950250
1-Bromo-2-iodobenzene 99% stabilised
1-Bromo-2-iodobenzene
Formel: C₆H₄BrI
MW: 282,91 g/mol
Kokpunkt: 120…121 °C (15 mmHg)
Smältpunkt: 9…10 °C
Densitet: 2,203 g/cm³
Flampunkt: >110 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001030
CAS nummer: 583-55-1
EINECS: 209-508-9
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.6620 to 1.6650 (20°C, 589 nm)
GC ≥98.5 %
Stabilizer Copper