1-Iodo-2-methylpropane 97% stabilised
Leverantör: Thermo Scientific
Fara
Synonyms: Isobutyliodide
223740250 223741000 223745000
ACRO223740250EA 1275 SEK
ACRO223740250 ACRO223741000 ACRO223745000
1-Iodo-2-methylpropane 97% stabilised
1-Iodo-2-methylpropane
Formel: C₄H₉I
MW: 184,02 g/mol
Kokpunkt: 120…121 °C (760 mmHg)
Smältpunkt: –93 °C
Densitet: 1,59 g/cm³
Flampunkt: 12 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001084
CAS nummer: 513-38-2
EINECS: 208-160-5
UN: 2391
ADR: 3,II
Merck Index: 13,05160
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer copper