(±)-Äppelsyra ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: (±)-Äppelsyra
A17874.0E A17874.30 A17874.36
A17874.0EEA 1650 SEK
A17874.0E A17874.30 A17874.36
(±)-Äppelsyra ≥98%
DL-Äppelsyra
Purity: 98% CAS No.: 6915-15-7 Molecular Formula: C4H6O5 Formula Weight: 134.09 Flash Point: 203°(397°F) Physical Form: Powder Density: 1.609 Appearance: White to pale cream Melting Point: 130-133° Solubility: Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene. Beilstein Reference: 1723539 Merck Reference: 14,5707 MDL No.: MFCD00064212
Formel: HOOCCH(OH)CH₂COOH
MW: 134,09 g/mol
Kokpunkt: 306 °C (1013 hPa)
Smältpunkt: 130…133 °C
Densitet: 1,609
Flampunkt: 203 °C (397 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00064212
CAS nummer: 617-48-1
EINECS: 230-022-8
Merck Index: 14,5707
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=57a7a54be23d2812f9b1a79aa7aaec92b4fde376&prid=web&clid=SAPDAM