DL-Metionin ≥99%
Leverantör: Thermo Scientific
Synonyms: 2-Amino-4-(methylthio)butanoic acid, (±)-2-Amino-4-(methylthio)butyric acid, Methioninum racemicum, Methionine racematic, H-DL-Met-OH, (±)-Methionine, (±)-2-Amino-4-(methylthio)butanoic acid
A11457.0B A11457.0I A11457.30
A11457.0BEA 2000 SEK
A11457.0B A11457.0I A11457.30
DL-Metionin ≥99%
DL-Metionin
Purity: 99% CAS No.: 59-51-8 Molecular Formula: C5H11NO2S Formula Weight: 149.21 Melting Point: ca 272° dec. Beilstein Reference: 636185 Merck Reference: 14,5975 MDL No.: MFCD00063096
Formel: C₅H₁₁NO₂S
MW: 149,21 g/mol
Smältpunkt: ∼ 272 °C (dec.)
Densitet: 1,34
Storage Temperature: Omgivande
MDL Number: MFCD00063096
CAS nummer: 59-51-8
EINECS: 200-432-1
Merck Index: 14,5975