1-Jodpropan 99% stabilised
Leverantör: Thermo Scientific
Varning
Synonyms: 1-Propyl iodide
174050500 174052500 174050010
ACRO174050500EA 620 SEK
ACRO174050500 ACRO174052500 ACRO174050010
1-Jodpropan 99% stabilised
1-Jodpropan
Formel: C₃H₇I
MW: 169,99 g/mol
Kokpunkt: 103 °C (1013 hPa)
Smältpunkt: –101 °C
Densitet: 1,74 g/cm³
Flampunkt: 44 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001095
CAS nummer: 107-08-4
EINECS: 203-460-2
UN: 2392
ADR: 3,III
Merck Index: 12,08047
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5060 (20°C, 589 nm)
GC ≤1.5 % (Sum of all by-products)
Stabilizer Copper