(±)-Linalool ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: (±)-3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethylocta-1,6-dien-3-ol
L0048-25ML L0048-500ML
TCIAL0048-25MLEA 215 SEK
TCIAL0048-25ML TCIAL0048-500ML
(±)-Linalool ≥96.0%
(±)-Linalool
Formel: C₁₀H₁₈O
MW: 154,25 g/mol
Kokpunkt: 200°C
Densitet: 0,87 g/cm³ (20 °C)
Flampunkt: 71°C
Storage Temperature: Omgivande
MDL Number: MFCD00008906
CAS nummer: 78-70-6
EINECS: 201-134-4
Merck Index: 14,05495
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5733b7430eaceba1185702c1393a094e38d967ab&vid=5733b7430eaceba1185702c1393a094e38d967ab&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %