Tween® 60 (Polysorbate), EMPROVE® ESSENTIAL JPE, Ph. Eur., NF suitable for use as excipient, SAFC®
Leverantör: MERCK PRODUCTION CHEMICALS
Synonyms: Polyoxyethylene sorbitan monostearate
8.17076.2500 8.17076.9050
8.17076.2500EA 5775 SEK
8.17076.2500 8.17076.9050
Tween® 60 (Polysorbate), EMPROVE® ESSENTIAL JPE, Ph. Eur., NF suitable for use as excipient, SAFC®
Tween® 60 (Polysorbate)
Formel: C₆₄H₁₂₆O₂₆
Kokpunkt: ∼110 °C (1013 hPa)
Densitet: 1,08 g/cm³ (20 °C)
Flampunkt: >149 °C
Storage Temperature: Omgivande
MDL Number: MFCD01780110
CAS nummer: 9005-67-8
EINECS: 500-020-4