Polysorbat 80, GPR RECTAPUR®
Leverantör: VWR Chemicals
Synonyms: Sorbitanmonooleat, etoxylerad
28830.291EA 1100 SEK
28830.291 28830.260 28830.320 28830.460
Polysorbat 80, GPR RECTAPUR®
Polysorbat 80
Kokpunkt: >100 °C (1013 hPa)
Smältpunkt: –21 °C
Densitet: 1,062 g/cm³ (20 °C)
Flampunkt: 149 °C (closed cup)
Storage Temperature: Omgivande
MDL Number: MFCD00082107
CAS nummer: 9005-65-6
EINECS: 500-019-9
Merck Index: 13,07664
Order Now
Specifikation testresultat
Identification Passes test
Density (20/4) 1.070 - 1.090
Hydroxyl value 65 - 80
Saponification value 45 - 55
Acid value Max 2 mg KOH/g
Water content Max 3%