TRITON™ X-100 non-ionic detergent, Millipore®
Leverantör: Merck Millipore Calbiochem
Fara
648462-1 648462-3
648462-1EA 2825 SEK
648462-1 648462-3
TRITON™ X-100 non-ionic detergent, Millipore®
TRITON™ X-100
Non-ionic detergent and emulsifier.
Kokpunkt: ∼200 °C (1013 hPa)
Smältpunkt: 6 °C
Densitet: 1,06 g/cm³ (20 °C)
Flampunkt: 251 °C (closed cup)
Storage Temperature: Kylskåp
MDL Number: MFCD00132505
CAS nummer: 9002-93-1
UN: 3082
ADR: 9,III
Merck Index: 13,06793